Learn More
Genscript Corporation CD44 hFc Chimera Human
Catalog No. 5000059076
Encompass
CD44 is a hyaluronan binding cell surface signal transducing receptor that influences motility cell survival and proliferation as well as the formation of tumor microenvironment CD44 contains two variable regions encoded by variable exons Alternative splicing which is often deregulated in cancer can produce various isoforms of CD44 with properties that may have different tissue specific effects and therefore even diverse effects on cancer progression
Catalog No. 50-000-59076 Supplier Genscript Corporation Supplier No. Z04209500
CD44 is a hyaluronan binding cell surface signal transducing receptor that influences motility cell survival and proliferation as well as the formation of tumor microenvironment CD44 contains two variable regions encoded by variable exons Alternative splicing which is often deregulated in cancer can produce various isoforms of CD44 with properties that may have different tissue specific effects and therefore even diverse effects on cancer progression