Learn More
Medchemexpress LLC CD44 Human HEK293 100ug
SDP
Catalog No. 5000184151
Encompass_Preferred
Recombinant human CD44 ectodomain expressed in HEK293 cells and supplied with a C-terminal 6xHis tag. The lyophilized protein is >95% pure by reducing SDS-PAGE and is provided with supporting datasheet, COA, and handling documentation. It is intended for research use in studies of cell adhesion, migration, and CD44-mediated signaling.
  • Expression in HEK293 cells for human-like post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Sequence length 200 amino acids; expected molecular weight 38-50 kDa.
Catalog No. 50-001-84151 Supplier Medchemexpress LLC Supplier No. HYP70507100UG
Recombinant human CD44 ectodomain expressed in HEK293 cells and supplied with a C-terminal 6xHis tag. The lyophilized protein is >95% pure by reducing SDS-PAGE and is provided with supporting datasheet, COA, and handling documentation. It is intended for research use in studies of cell adhesion, migration, and CD44-mediated signaling.
  • Expression in HEK293 cells for human-like post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • Sequence length 200 amino acids; expected molecular weight 38-50 kDa.