Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CD44 Human HEK293 5ug
SDP

Catalog No. 5000184811
Encompass_Preferred

Recombinant human CD44 extracellular domain produced in HEK293 cells and supplied as a C-terminal His-tagged, lyophilized protein. It is formulated for reconstitution and validated for biochemical and cell-based assays, offering high purity and characterized biological activity for binding and proliferation studies.

  • Expressed in human HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Amino acid range Q21-P220 representing the extracellular domain.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Do not reconstitute below 100 μg/mL in ddH2O.
  • Store at -20°C; freeze aliquots for long-term storage at -80°C.
  • Validated in binding and cell proliferation assays with reported activity.

Catalog No. 50-001-84811 Supplier Medchemexpress LLC Supplier No. HYP705075UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human CD44 extracellular domain produced in HEK293 cells and supplied as a C-terminal His-tagged, lyophilized protein. It is formulated for reconstitution and validated for biochemical and cell-based assays, offering high purity and characterized biological activity for binding and proliferation studies.

  • Expressed in human HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Amino acid range Q21-P220 representing the extracellular domain.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Do not reconstitute below 100 μg/mL in ddH2O.
  • Store at -20°C; freeze aliquots for long-term storage at -80°C.
  • Validated in binding and cell proliferation assays with reported activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.