Learn More
Bioss CD44 - Hyaluron Binding Region
SDP
Catalog No. 5000066070
Encompass_Preferred
CD44 (Hyaluron Binding Region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications ELISA M591Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-66070 Supplier Bioss Supplier No. BSM70399M
CD44 (Hyaluron Binding Region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications ELISA M591Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol