Learn More
Medchemexpress LLC CD44 Mouse HEK293 10ug
SDP
Catalog No. 5000178930
Encompass_Preferred
The CD44 protein binds to hyaluronic acid and TGF- receptors and contributes to cytokine function CD44 is involved in T cell activation protein phosphorylation and Wnt signaling and is strategically located in the plasma membrane and microvilli CD44 Protein Mouse (HEK293 His) is the recombinant mouse-derived CD44 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78930 Supplier Medchemexpress LLC Supplier No. HYP7429110UG
The CD44 protein binds to hyaluronic acid and TGF- receptors and contributes to cytokine function CD44 is involved in T cell activation protein phosphorylation and Wnt signaling and is strategically located in the plasma membrane and microvilli CD44 Protein Mouse (HEK293 His) is the recombinant mouse-derived CD44 protein expressed by HEK293 with C-His labeled tag