Learn More
Enzo Life Sciences CD44 std (human) monoclonal antibody (SFF-304) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX801089C100
Encompass_Preferred
Clone: SFF-304. Isotype: IgG1. Immunogen: Recombinant human CD44. Species reactivity: Human. Specificity: Does not distinguish between CD44 splice variants. Applications: Flow Cytometry, IHC (FS), IHC (PS), WB. Formulation: Liquid. In PBS, pH 7.2-7.4.
Catalog No. 50-200-9718