Learn More
ACROBiosystems Recombinant Protein;Human CD44 Protein, Fc Tag (HPLC-verified);HEK293;1MG;PG1,CD44,CDW44,CSPG8,ECMR-III,HCELL,HUTCH-I,IN,LHR,MC56,MDU2,MDU3,MIC4,Pgp1,Epican,Pgp-1
Catalog No. 501658637
Encompass_Preferred
Recombinant Protein;Human CD44 Protein, Fc Tag (HPLC-verified);1MG;Human CD44, Fc Tag (PG1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession # AAH04372).;This protein carries a human IgG1 Fc tag at the C-terminus, and has a calculated MW of 48.7 kDa. The predicted N-terminus is Gln 21. The reducing (R) protein migrates as 76 kDa in SDS-PAGE due to glycosylation.
Catalog No. 50-165-8637 Supplier ACROBiosystems Supplier No. PG1H52551MG
Recombinant Protein;Human CD44 Protein, Fc Tag (HPLC-verified);1MG;Human CD44, Fc Tag (PG1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession # AAH04372).;This protein carries a human IgG1 Fc tag at the C-terminus, and has a calculated MW of 48.7 kDa. The predicted N-terminus is Gln 21. The reducing (R) protein migrates as 76 kDa in SDS-PAGE due to glycosylation.