Learn More
ACROBiosystems Recombinant Protein;Human CD44 Protein;HEK293;100UG;CD4,CD44,CDW44,CSPG8,ECMR-III,HCELL,HUTCH-I,IN,LHR,MC56,MDU2,MDU3,MIC4,Pgp1,Epican,Pgp-1
Catalog No. 501658048
Encompass_Preferred
Recombinant Protein;Human CD44 Protein;100UG;Human CD44, His Tag (CD4-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession # AAH04372).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.9 kDa. The protein migrates as 35-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8048 Supplier ACROBiosystems Supplier No. CD4H5226100UG
Recombinant Protein;Human CD44 Protein;100UG;Human CD44, His Tag (CD4-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession # AAH04372).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.9 kDa. The protein migrates as 35-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.