Learn More
Enzo Life Sciences CD44v4 Recombinant monoclonal antibody (DC44v4/1700R) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3490100
Encompass_Preferred
Clone: CD44v4/1700R. Isotype: IgGκ. Immunogen: An amino acid sequence from the variant 4 domain of CD44. Species reactivity: Human. Specificity: This antibody recognizes an epitope encoded by exon v4 on the variant portion of human CD44. Applications: Flow Cytometry, IF, IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2879