Learn More
Eurofins Discoverx CD45, 250 µg. (14-618M)
Supplier:  Eurofins Discoverx 14618M
Encompass
The CD45 product is purified N terminal 6xHis-tagged recombinant CD45 enzyme. This enzyme contains amino acids 598-end. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 0.829 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9064