Learn More
Fluidigm Corp CD45-89Y(CLONE:HI30)
SDP
Supplier:  Fluidigm Corp 3089003B
Encompass
CD45, also known as leukocyte common antigen (LCA) and T200, is a type I transmembrane glycoprotein. It is expressed on the plasma membrane of all hematopoietic cells except mature red blood cells and platelets. Its intracellular domain is a tyrosine phosphatase that serves to regulate signal transduction in most hematopoietic cells.
Catalog No. NC0782474