Learn More
CAPRICO BIOTECHNOLOGIES INC CD45 Antibody, APC Conjugated
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4016045
Encompass_Preferred
The Caprico CD45 Antibody APC Conjugated is a mouse monoclonal antibody that specifically binds to the CD45 antigen, also known as the leukocyte common antigen. It is conjugated to the fluorescent dye APC (allophycocyanin) and is designed for use in flow cytometry, immunohistochemistry, and immunofluorescence assays. This antibody can be used to identify and analyze leukocytes and hematopoietic cells in human, non-human primate, canine, and rat samples. It is available in 100 tests of 50μl volume at a concentration of 20μg/ml.
Catalog No. 50-227-9166