Learn More
CAPRICO BIOTECHNOLOGIES INC CD45 APC-Cyanine7 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4016095
Encompass_Preferred
The CD45 APC-Cyanine7 Antibody from Caprico Biosciences is a monoclonal antibody that specifically binds to CD45, also known as the leukocyte common antigen. It is conjugated to APC-Cyanine7 and is suitable for flow cytometry applications. This antibody recognizes all isoforms of CD45 and can be used to identify and analyze leukocytes. It is recommended for use in human samples. The APC-Cyanine7 fluorochrome has an excitation max of 633nm and emission max of 760nm. This antibody is available in sizes ranging from 25ug to 1mg.
Catalog No. 50-227-9167