Learn More
Enzo Life Sciences CD45 (human) clonal antibody (E19-G) (Ready-to-Use) (15ml)
SDP
Supplier:  Enzo Life Sciences ALX810410L015
Encompass_Preferred
Clone: E19-G. Isotype: IgG. Immunogen: Synthetic peptide corresponding to a C-terminal sequence of human CD45. Species reactivity: Human. Applications: IHC (PS). Purity: Major clone of IgGs obtained from immunoaffinity purified immunoglobulins corresponding to immunogenic peptide. Formulation: Liquid. Ready-to-use solution in 20mM TRIS/HCl, pH 8.0, containing 20mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-0088