Learn More
STEMCELL Technologies Anti-Human CD45 Antibody, Clone HI30, APC, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60018AZ.1
Encompass
Clone HI30 is a mouse monoclonal antibody that reacts with human and chimpanzee CD45 a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes platelets and plasma cells. Verified for use with flow cytometry and cell separation assays use this antibody in applications requiring the detection of CD45. CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes. This antibody clone has also been verified for purity assessments of cells isolated with EasySep kits including EasySep Human T Cell Enrichment Kit (Catalog 19051) EasySep Human CD4 T Cell Enrichment Kit (Catalog 19052) and EasySep Human CD45 Depletion Kit (Catalog 18259 partial blocking may be observed). Visit STEMCELL.com for more information and related products.
Catalog No. 50-239-0417