Learn More
CAPRICO BIOTECHNOLOGIES INC CD45 MFLUOR510 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4016215
Encompass_Preferred
The Caprico CD45 MFLUOR510 Antibody is a mouse monoclonal antibody that recognizes human CD45. It is conjugated to MFLUOR510, a bright, photostable fluorescent dye with excitation/emission of 506/528 nm. This antibody can be used for flow cytometry, immunofluorescence microscopy, and immunohistochemistry assays. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. The immunogen is a recombinant human CD45 protein. This antibody recognizes all isoforms of CD45 and has been tested on human peripheral blood samples. It is recommended for detecting CD45 expression on leukocytes.
Catalog No. 50-227-9178