Learn More
Medchemexpress LLC CD45 Mouse HEK293 10ug
SDP
Catalog No. 5000182912
Encompass_Preferred
The CD45 protein is a multifunctional member of the protein tyrosine phosphatase family with heparan sulfate proteoglycan and heparin binding activity as well as protein tyrosine phosphatase activity It plays a crucial role in lymphocyte differentiation macromolecular metabolic processes and signal transduction regulation CD45 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived CD45 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-82912 Supplier Medchemexpress LLC Supplier No. HYP7291610UG
The CD45 protein is a multifunctional member of the protein tyrosine phosphatase family with heparan sulfate proteoglycan and heparin binding activity as well as protein tyrosine phosphatase activity It plays a crucial role in lymphocyte differentiation macromolecular metabolic processes and signal transduction regulation CD45 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived CD45 protein expressed by HEK293 with C-hFc labeled tag