Learn More
CAPRICO BIOTECHNOLOGIES INC CD45 PE-Cyanine5 Antibody, Clone HI30, 100 Tests, 100 μL
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4016075
Encompass_Preferred
The CD45 PE-Cyanine5 Antibody from Caprico Biotechnologies is a mouse monoclonal antibody that recognizes human CD45. It is conjugated to PE-Cyanine5 and is intended for use in flow cytometry. This antibody clone is HI30. It is supplied as 100 tests in a volume of 100 μL. The immunogen is a cell preparation - human: T-cell leukemia line CEM. This antibody can be used for immunophenotyping of leukocytes and is recommended for use with human peripheral blood cells.
Catalog No. 50-227-9171