Learn More
CAPRICO BIOTECHNOLOGIES INC CD45 PE-Cyanine7 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4016085
Encompass_Preferred
The CD45 PE-Cyanine7 Antibody from Caprico Biotechnologies is a mouse monoclonal antibody that recognizes human CD45. It is conjugated to PE-Cyanine7 and is suitable for flow cytometry applications. This antibody can be used to identify leukocytes and hematopoietic cells. It specifically binds to all isoforms of the CD45 antigen. The clone for this antibody is HI30. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. The concentration is 0.5 mg/mL.
Catalog No. 50-227-9170