Learn More
CAPRICO BIOTECHNOLOGIES INC CD45 PerCP Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4016035
Encompass_Preferred
The Caprico CD45 PerCP Antibody is a mouse monoclonal antibody that specifically binds to human CD45. It is conjugated to PerCP and is intended for flow cytometry applications. This antibody recognizes all isoforms of CD45 and has been tested on human peripheral blood samples. It can be used to identify leukocytes and hematopoietic cells. The antibody is supplied as a liquid in PBS with 0.1% sodium azide and 2 mg/mL BSA. It has a concentration of 0.2 mg/mL and is available in sizes of 100 tests and 1000 tests.
Catalog No. 50-227-9172