Learn More
Sino Biological Human CD45-AB Protein (His Tag) 100ug
Supplier:  Sino Biological 16884H08H100
Encompass_Preferred
A DNA sequence encoding the human PTPRC (XP_006711535.1) (Met1-Lys529) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4769