Learn More
Sino Biological Human CD45 Protein (ECD, His Tag) 50ug
Supplier:  Sino Biological 14197H08H50
Encompass_Preferred
A DNA sequence encoding the human CD45 (NP_002829.3) (Gln26-Lys575) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5582