Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5788MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 158-4D3. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: Daudi, Raji, IM9, human lymphocytes. Human lymph nodes, tonsils. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Stimulated human leukocytes. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 205-220kDa.Chromosome: 1q31.3. Gene Symbol: PTPRC. Gene ID: 5788. Ensemble: ENSG00000262418. Uniprot: NP_002829.
Catalog No. 50-188-5110