Learn More
CAPRICO BIOTECHNOLOGIES INC CD45 APC-Cyanine7 (F10-89-4) RUO
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4087095
Encompass_Preferred
CD45 APC-Cyanine7 is a monoclonal antibody conjugated to APC-Cyanine7 dye, used to identify leukocyte populations in flow cytometry. It binds to all isoforms of CD45, a transmembrane protein tyrosine phosphatase expressed on all hematopoietic cells except erythrocytes and platelets. This antibody is intended for research use only, not for use in diagnostic or therapeutic procedures. It is optimized for flow cytometry analysis of human peripheral blood cells.
Catalog No. 50-227-9184