Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5788MSM13P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: DF-B1. Clonality: Monoclonal. Host: Mouse. Isotype: IgG3 kappa. Positive Control: Ramos, U-698, or GA-10 cells. Tonsil. Cellular Staining: Cell surface, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Extract from Alexander.’s hepatoma cell line L428 (Workshop V). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 180-220kDa.Chromosome: 1q31.3. Gene Symbol: PTPRC. Gene ID: 5788. Ensemble: ENSG00000262418. Uniprot: NP_002829.
Catalog No. 50-188-5098