Learn More
Bioss CD46 - Extracellular region An
SDP
Catalog No. 5000067038
Encompass_Preferred
CD46 (Extracellular region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M037Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-67038 Supplier Bioss Supplier No. BSM70400M
CD46 (Extracellular region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M037Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol