Learn More
Genscript Corporation CD46 His Cynomolgus
Catalog No. 5000058994
Encompass
CD46 was discovered in 1986 during a search for novel C3b-binding proteins CD46 is expressed ubiquitously and functions as a co-factor in the factor I-mediated proteolytic cleavage of C3b and C4b Its vital role in preventing complement deposition on host tissue is underpinned by the fact that deficiency of CD46 is a predisposing factor for numerous disease conditions arising from complement-mediated self-attack
Catalog No. 50-000-58994 Supplier Genscript Corporation Supplier No. Z04123500
CD46 was discovered in 1986 during a search for novel C3b-binding proteins CD46 is expressed ubiquitously and functions as a co-factor in the factor I-mediated proteolytic cleavage of C3b and C4b Its vital role in preventing complement deposition on host tissue is underpinned by the fact that deficiency of CD46 is a predisposing factor for numerous disease conditions arising from complement-mediated self-attack