Learn More
ACROBiosystems ACROBiosystems Human CD46 Protein, His Tag
Supplier:  ACROBiosystems CD6H52261MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD46, His Tag (CD6-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 35 - Asp 328 (Accession # AAH30594.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.6 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7491