Learn More
ACROBiosystems Recombinant Protein;Human CD46 Protein;HEK293;50UG;CD6,CD46,AHUS2,MCP,MIC10,TLX,TRA2.10
Catalog No. 501657851
Encompass_Preferred
Recombinant Protein;Human CD46 Protein;50UG;Human CD46, His Tag (CD6-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 35 - Asp 328 (Accession # AAH30594.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.6 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7851 Supplier ACROBiosystems Supplier No. CD6H522650UG
Recombinant Protein;Human CD46 Protein;50UG;Human CD46, His Tag (CD6-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 35 - Asp 328 (Accession # AAH30594.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.6 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.