Learn More
Sino Biological Anti-CD47 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12283MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD47 (rh CD47; Catalog#12283-HCCH; NP_942088.1; Met1-Pro139). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-222-2298