Learn More
Bioss CD47 - Extracellular region An
SDP
Catalog No. 5000065292
Encompass_Preferred
CD47 (Extracellular region) Antibody Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M054Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65292 Supplier Bioss Supplier No. BSM70414M
CD47 (Extracellular region) Antibody Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M054Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol