Learn More
ACROBiosystems ACROBiosystems Rat CD47 Protein, Fc Tag
Supplier:  ACROBiosystems CD7R5256100UG
Encompass_Preferred
Recombinant Protein;100UG;Rat CD47, Fc Tag (CD7-R5256) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # P97829).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. As a result of glycosylation, the protein migrates as 50-66 kDa under reducing (R) condition, and 100-130 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag (Cat. No. SIA-M5252) at 10 μg/mL (100 μL/well) can bind Rat CD47, Fc Tag (Cat. No. CD7-R5256) with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-195-7368