Learn More
ACROBiosystems Rabbit CD47 Protein, Fc Tag, Lyophilized, 1 mg
Catalog No. 501957414
Encompass_Preferred
Rabbit CD47 protein is a leukocyte surface antigen expressed from human HEK293 cells. It contains the extracellular domain of CD47 fused with a human IgG1 Fc tag at the C-terminus. CD47 functions as an adhesion receptor for THBS1 on platelets and acts as a receptor for SIRPA, playing a critical role in modulating integrins, cell adhesion, memory formation, and synaptic plasticity.
  • Purity >95% as determined by SDS-PAGE
  • Calculated molecular weight of 40.4 kDa
  • Migrates as 48-58 kDa on SDS-PAGE under reducing conditions due to glycosylation
  • Lyophilized from a 0.22 μm filtered solution in Tris, glycine, arginine, and NaCl, pH 7.5, with trehalose
  • Endotoxin level <1.0 EU per μg by the LAL method
  • Stable for 12 months at -20°C to -70°C in lyophilized state
Catalog No. 50-195-7414 Supplier ACROBiosystems Supplier No. CD7R52571MG
Rabbit CD47 protein is a leukocyte surface antigen expressed from human HEK293 cells. It contains the extracellular domain of CD47 fused with a human IgG1 Fc tag at the C-terminus. CD47 functions as an adhesion receptor for THBS1 on platelets and acts as a receptor for SIRPA, playing a critical role in modulating integrins, cell adhesion, memory formation, and synaptic plasticity.
  • Purity >95% as determined by SDS-PAGE
  • Calculated molecular weight of 40.4 kDa
  • Migrates as 48-58 kDa on SDS-PAGE under reducing conditions due to glycosylation
  • Lyophilized from a 0.22 μm filtered solution in Tris, glycine, arginine, and NaCl, pH 7.5, with trehalose
  • Endotoxin level <1.0 EU per μg by the LAL method
  • Stable for 12 months at -20°C to -70°C in lyophilized state