Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque CD47 Protein, Fc Tag
Supplier:  ACROBiosystems CD7C52521MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque CD47, Fc Tag (CD7-C5252) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Accession # F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. As a result of glycosylation, the protein migrates as 50-66 kDa under reducing (R) condition, and 100-130 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag (Cat. No. SIA-M5252) at 10 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque CD47, Fc Tag (Cat. No. CD7-C5252) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-195-7528