Recombinant Protein;200UG;MABSol Biotinylated Human CD47, Fc,Avitag (CD7-H82F6) is expressed from human HEK293 cells. It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 42.2 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, His Tag (HPLC-verified) (Cat. No. SIA-H5225) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD47, Fc,Avitag (Cat. No. CD7-H82F6) with a linear range of 0.008-0.25 μg/mL (QC tested).;Immobilized Biotinylated Human CD47, Fc,Avitag (Cat. No. CD7-H82F6) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.2 μg/well) plate, can bind Cynomolgus SIRP alpha, His Tag (Cat. No. SIA-C52H7) with a linear range of 0.039-0.625 μg/mL (Routinely tested).