Learn More
Genscript Corporation CD47 hFc ChimeraBiotin Avi
Catalog No. 5000056865
Encompass
CD47 (Cluster of Differentiation 47) also known as integrin associated protein (IAP) is a transmembrane protein that in humans is encoded by the CD47 gene CD47 belongs to the immunoglobulin superfamily and partners with membrane integrins and also binds the ligands thrombospondin-1 (TSP-1) and signal-regulatory protein alpha (SIRP ) CD-47 acts as a don t eat me signal to macrophages of the immune system which has made it a potential therapeutic target in some cancers
Catalog No. 50-000-56865 Supplier Genscript Corporation Supplier No. Z04069500
CD47 (Cluster of Differentiation 47) also known as integrin associated protein (IAP) is a transmembrane protein that in humans is encoded by the CD47 gene CD47 belongs to the immunoglobulin superfamily and partners with membrane integrins and also binds the ligands thrombospondin-1 (TSP-1) and signal-regulatory protein alpha (SIRP ) CD-47 acts as a don t eat me signal to macrophages of the immune system which has made it a potential therapeutic target in some cancers