Learn More
ACROBiosystems ACROBiosystems Canine CD47 Protein, His Tag
Supplier: ACROBiosystems CD7C52H5100UG
Recombinant Protein;100UG;Canine CD47, His Tag (CD7-C52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 140 (Accession # F1P6D7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.8 kDa. The protein migrates as 27-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Fc Tag (HPLC-verified) (Cat. No. SIA-H5251) at 5 μg/mL (100 μL/well) can bind Canine CD47, His Tag (Cat. No. CD7-C52H5) with a linear range of 5-78 ng/mL (QC tested).