Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse CD47 Protein, His,Avitag™
Supplier:  ACROBiosystems CD7M82E4200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse CD47, His,Avitag (CD7-M82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.4 kDa. The protein migrates as 30-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse CD47, His,Avitag (Cat. No. CD7-M82E4) at 2 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SIA-M5252) with a linear range of 0.019-0.625 μg/mL (QC tested).
Catalog No. 50-195-6687