Learn More
Medchemexpress LLC HY-P7336 10ug Medchemexpress, CD47, His (HEK293-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 502083149
Encompass_Preferred
Medchemexpress, HY-P7336 10ug CD47, His (HEK293-expressed), Human CAS: Recombinant Human CD47, His (HEK293-expressed) is a polypeptide chain with His tag produced in HEK293 cells. CD47 is a prominent target in cancer immunotherapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3149 Supplier Medchemexpress LLC Supplier No. HYP733610UG
Medchemexpress, HY-P7336 10ug CD47, His (HEK293-expressed), Human CAS: Recombinant Human CD47, His (HEK293-expressed) is a polypeptide chain with His tag produced in HEK293 cells. CD47 is a prominent target in cancer immunotherapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.