Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Mouse CD47 Protein, Mouse IgG2a Fc Tag, Low Endotoxin, 1 mg

Catalog No. 501957206
Encompass_Preferred

This mouse CD47 protein is expressed from human 293 cells (HEK293) and contains amino acids Gln 19 to Lys 140. It is a leukocyte surface antigen that plays key roles in cell adhesion and modulation of integrins, serving as the receptor for SIRPA. This product is provided in a lyophilized state and features a mouse IgG2a Fc tag at the C-terminus.

  • Expressed in human HEK293 cells
  • Contains one Ig-like V-type domain
  • Purity >95% by SDS-PAGE and >90% by SEC-MALS
  • Endotoxin level <0.01 EU per μg
  • Calculated molecular weight of 40.7 kDa
  • Lyophilized from a 0.22 μm filtered solution with trehalose

Catalog No. 50-195-7206 Supplier ACROBiosystems Supplier No. CD7M525A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This mouse CD47 protein is expressed from human 293 cells (HEK293) and contains amino acids Gln 19 to Lys 140. It is a leukocyte surface antigen that plays key roles in cell adhesion and modulation of integrins, serving as the receptor for SIRPA. This product is provided in a lyophilized state and features a mouse IgG2a Fc tag at the C-terminus.

  • Expressed in human HEK293 cells
  • Contains one Ig-like V-type domain
  • Purity >95% by SDS-PAGE and >90% by SEC-MALS
  • Endotoxin level <0.01 EU per μg
  • Calculated molecular weight of 40.7 kDa
  • Lyophilized from a 0.22 μm filtered solution with trehalose

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.