Learn More
Sino Biological Human CD47 Protein (His & AVI Tag), Biotinylated 100ug
Catalog No. 502115341
Encompass_Preferred
A DNA sequence encoding the human CD47 (NP_942088.1) (Met1-Pro139) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-211-5341 Supplier Sino Biological Supplier No. 12283H27HB100
A DNA sequence encoding the human CD47 (NP_942088.1) (Met1-Pro139) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.