Learn More
Sino Biological Human CD47 Protein (ECD,His Tag) 1mg
Catalog No. 502115498
Encompass_Preferred
A DNA sequence encoding the human CD47 (NP_942088.1) (Met1-Pro139) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5498 Supplier Sino Biological Supplier No. 12283H08H1
A DNA sequence encoding the human CD47 (NP_942088.1) (Met1-Pro139) was expressed with a polyhistidine tag at the C-terminus.