Learn More
Sino Biological Recombinant Human CD47 Protein (ECD, His Tag), HPLC-verified, High activity
Supplier:  Sino Biological 12283H08H100
Encompass_Preferred
A DNA sequence encoding the human CD47 (NP_942088.1) (Gln19-Pro139) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6293