Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CD47 protein, rat (HEK293, Fc) | 95.0% | 1 MG
SDP

Catalog No. 5000293331
Encompass_Preferred

Recombinant rat CD47 extracellular domain produced in HEK293 cells and presented as a C-terminal human Fc fusion protein. Intended for research use in in vitro binding studies, biochemical characterization, and assay development. Supplied purified and frozen to preserve structural integrity and activity.

  • Recombinant rat CD47 extracellular domain.
  • Expressed in HEK293 mammalian cells.
  • C-terminal human Fc fusion tag for stability and detection.
  • Purity approximately 95.0%.
  • Supplied frozen; store at -20°C.
  • Available in a 1 mg format.

Catalog No. 50-002-93331 Supplier Medchemexpress LLC Supplier No. HYP754021MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat CD47 extracellular domain produced in HEK293 cells and presented as a C-terminal human Fc fusion protein. Intended for research use in in vitro binding studies, biochemical characterization, and assay development. Supplied purified and frozen to preserve structural integrity and activity.

  • Recombinant rat CD47 extracellular domain.
  • Expressed in HEK293 mammalian cells.
  • C-terminal human Fc fusion tag for stability and detection.
  • Purity approximately 95.0%.
  • Supplied frozen; store at -20°C.
  • Available in a 1 mg format.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.