Learn More
Medchemexpress LLC CD47 protein, rat (HEK293, Fc) | 95.0% | 100 UG
SDP
Catalog No. 5000291783
Encompass_Preferred
Recombinant rat CD47 (leukocyte surface antigen) produced in HEK293 cells with a C-terminal human Fc fusion. Supplied as 100 μg for research applications; store at -20°C and ship on blue ice. Protein accession P97829 and gene ID 29364 are provided where available.
Catalog No. 50-002-91783 Supplier Medchemexpress LLC Supplier No. HYP75402100UG
Recombinant rat CD47 (leukocyte surface antigen) produced in HEK293 cells with a C-terminal human Fc fusion. Supplied as 100 μg for research applications; store at -20°C and ship on blue ice. Protein accession P97829 and gene ID 29364 are provided where available.