Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CD47 protein, rat (HEK293, His) | >95.0% | 10 UG
SDP

Catalog No. 5000293008
Encompass_Preferred

Recombinant rat CD47 extracellular domain expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized and intended for research use; the protein is highly pure and shows heterogeneous migration by SDS-PAGE due to glycosylation.

  • Recombinant extracellular domain (Q19-K140) with C-terminal His tag.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight 23-40 kDa because of glycosylation.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O; aliquot and store at -20°C or -80°C for long term.
  • Suitable for immunoassays, binding studies, and antibody characterization.

Catalog No. 50-002-93008 Supplier Medchemexpress LLC Supplier No. HYP7540110UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat CD47 extracellular domain expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized and intended for research use; the protein is highly pure and shows heterogeneous migration by SDS-PAGE due to glycosylation.

  • Recombinant extracellular domain (Q19-K140) with C-terminal His tag.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight 23-40 kDa because of glycosylation.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O; aliquot and store at -20°C or -80°C for long term.
  • Suitable for immunoassays, binding studies, and antibody characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.