Learn More
ACROBiosystems Recombinant Protein;Mouse CD47 Protein;HEK293;1MG;CD7,CD47,MER6,IAP,OA3
Supplier:  ACROBiosystems CD7M522B1MG
Encompass_Preferred
Recombinant Protein;Mouse CD47 Protein;1MG;Mouse CD47 (CD7-M522b) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 33-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse SIRP alpha, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. SIA-M5252) at 10 μg/mL (100 μL/well) can bind Mouse CD47 (Cat. No. CD7-M522b) with a linear range of 0.078-0.625 μg/mL (QC tested).
Catalog No. 50-165-8058