Learn More
ACROBiosystems Recombinant Protein;Mouse CD47 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;CD7,CD47,MER6,IAP,OA3
Supplier:  ACROBiosystems CD7M525A100UG
Encompass_Preferred
Recombinant Protein;Mouse CD47 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (CD7-M525a) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.7 kDa. The protein migrates as 55-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-M525a) at 10 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alpha, Fc Tag (Cat. No. CDA-H82F2) with a linear range of 0.16-5 μg/mL (QC tested).
Catalog No. 50-165-7932