Learn More
Aviva Systems Biology Corp CD48 Antibody - N-terminal region (ARP63333_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63333_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD48. It was validated on Western Blot. . Alternate names: BCM1, BLAST, BLAST1, MEM-102, SLAMF2, hCD48, mCD48ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-287-941