Learn More
Cayman Chemical CD48 Long Isoformhumnrec 100ug
SDP
Catalog No. 502948671
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged CD48 expressed in HEK293 cells • Amino acids: 27-220 • MW: 23.8 kDa
Catalog No. 50-294-8671 Supplier Cayman Chemical Supplier No. 31833100 UG
Source: Active recombinant human C-terminal His-tagged CD48 expressed in HEK293 cells • Amino acids: 27-220 • MW: 23.8 kDa